5-Acetyluracil ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Acetylpyrimidine-2,4(1H,3H)-dione

044053-500MG 044053-1G 044053-5G
101863-886EA 556.92 USD
101863-886 101863-888 101863-890
5-Acetyluracil ≥95%
5-Acetyluracil

Hazard: Irritant

Formula: C₆H₆N₂O₃
Melting Pt: 265 °C
MDL Number: MFCD00151968
CAS Number: 6214-65-9

Order Now

Specification Test Results

Melting point 265°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR