Methyl-3-[(hydroxyimino)methyl]-1H-indole-2-carboxylate ≥95%

Supplier: Matrix Scientific

043979-500MG 043979-1G 043979-5G
101863-494EA 278.46 USD
101863-494 101863-496 101863-498
Methyl-3-[(hydroxyimino)methyl]-1H-indole-2-carboxylate ≥95%
Methyl-3-[(hydroxyimino)methyl]-1H-indole-2-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₀N₂O₃
MW: 218.21 g/mol
Melting Pt: 206…208 °C
MDL Number: MFCD04125006
CAS Number: 861211-41-8

Order Now

Specification Test Results

Melting point 206-208°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR