2-Hydroxy-4'-(methylsulfonyl)acetophenone ≥95%

Supplier: Matrix Scientific

043708-500MG 043708-1G 043708-5G
101862-164EA 130.77 USD
101862-164 101862-166 101862-168
2-Hydroxy-4'-(methylsulfonyl)acetophenone ≥95%
2-Hydroxy-4'-(methylsulphonyl)acetophenone

Hazard: Irritant

Formula: C₉H₁₀O₄S
Melting Pt: 136…138 °C
CAS Number: 197240-27-0

Order Now

Specification Test Results

Melting point 136-138°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR