3-Bromo-7-nitro-1H-indazole ≥95% for synthesis

Supplier: Matrix Scientific

Synonyms: 3-Bromo-7-nitro-1H-indazole, 3-Bromo-7-nitroindazole

043703-1G 043703-5G
101862-144EA 35.38 USD
101862-144 101862-146
3-Bromo-7-nitro-1H-indazole ≥95% for synthesis
3-Bromo-7-nitro-1H-indazole

Hazard code(s): Irritant

Formula: C₇H₄BrN₃O₂
Melting Pt: 182 °C
MDL Number: MFCD00159910
CAS Number: 74209-34-0

Order Now

Specification Test Results

Melting point 182°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR