(2-Chloro-1,3-thiazol-5-yl)methyl phenyl ether ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-5-(phenoxymethyl)thiazole, 2-Chloro-5-(phenoxymethyl)-1,3-thiazole

043665-500MG 043665-1G 043665-5G
101861-958EA 181.54 USD
101861-958 101861-960 101861-962
(2-Chloro-1,3-thiazol-5-yl)methyl phenyl ether ≥95%
(2-Chloro-1,3-thiazol-5-yl)methyl phenyl ether

Hazard: Irritant

Formula: C₁₀H₈NOClS
Melting Pt: 49…51 °C
MDL Number: MFCD01443853
CAS Number: 339018-23-4

Order Now

Specification Test Results

Melting point 49-51°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR