5-Amino-2,4-difluorophenol ≥95%

Supplier: Matrix Scientific

Synonyms: 5-amino-2,4-difluorophenol

043566-500MG 043566-1G 043566-5G
101861-472EA 84.62 USD
101861-472 101861-474 101861-476
5-Amino-2,4-difluorophenol ≥95%
5-Amino-2,4-difluorophenol

Hazard: Irritant

Formula: C₆H₅F₂NO
Melting Pt: 189…190 °C
MDL Number: MFCD04973718
CAS Number: 113512-71-3
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 189-190°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR