Ethyl-2-[(2-chlorobenzoyl)amino]acetate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-[(2-chlorophenyl)formamido]acetate

043499-500MG 043499-1G 043499-5G
101861-122EA 181.54 USD
101861-122 101861-124 101861-126
Ethyl-2-[(2-chlorobenzoyl)amino]acetate ≥95%
Ethyl-2-[(2-chlorobenzoyl)amino]acetate

Hazard: Irritant

Formula: C₁₁H₁₂ClNO₃
Melting Pt: 75…77 °C
MDL Number: MFCD00793745
CAS Number: 66824-94-0

Order Now

Specification Test Results

Melting point 75-77°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR