Methyl-1-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-4-piperidinecarboxylate ≥95%

Supplier: Matrix Scientific

043463-500MG 043463-1G 043463-5G
101860-924EA 747.69 USD
101860-924 101860-926 101860-928
Methyl-1-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-4-piperidinecarboxylate ≥95%
Methyl-1-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-4-piperidinecarboxylate

Hazard: Irritant

Formula: C₁₃H₁₄ClF₃N₂O₂
Melting Pt: 52…54 °C
MDL Number: MFCD00172275
CAS Number: 321430-51-7

Order Now

Specification Test Results

Melting point 52-54°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR