5-(4-Methylpiperazino)-2-nitroaniline ≥95%

Supplier: Matrix Scientific

Synonyms: 5-(4-Methylpiperazin-1-yl)-2-nitroaniline

043297-500MG 043297-1G 043297-5G
101860-106EA 29.23 USD
101860-106 101860-108 101860-110
5-(4-Methylpiperazino)-2-nitroaniline ≥95%
5-(4-Methylpiperazino)-2-nitroaniline

Hazard: Irritant

Formula: C₁₁H₁₆N₄O₂
Melting Pt: 152…155 °C
MDL Number: MFCD00545964
CAS Number: 23491-48-7

Order Now

Specification Test Results

Melting point 152-155°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR