5-Amino-1-(tert-butyl)-3-(4-fluorophenyl)-1H-pyrazole-4-carbonitrile ≥95%

Supplier: Matrix Scientific

043237-500MG 043237-1G 043237-5G
101859-836EA 260 USD
101859-836 101859-838 101859-840
5-Amino-1-(tert-butyl)-3-(4-fluorophenyl)-1H-pyrazole-4-carbonitrile ≥95%
5-Amino-1-(tert-butyl)-3-(4-fluorophenyl)-1H-pyrazole-4-carbonitrile

Hazard: Irritant

Formula: C₁₄H₁₅FN₄
Melting Pt: 114…116 °C
MDL Number: MFCD01568265
CAS Number: 320422-34-2

Order Now

Specification Test Results

Melting point 114-116°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR