2-Methyl-1,3-benzoxazol-4-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Amino-2-methyl-1,3-benzoxazole

043181-500MG 043181-1G 043181-5G
101859-544EA 153.85 USD
101859-544 101859-546 101859-548
2-Methyl-1,3-benzoxazol-4-amine ≥95%
2-Methyl-1,3-benzoxazol-4-amine

Hazard: Irritant

Formula: C₁₁H₁₄N₂O
Melting Pt: 76…78 °C
CAS Number: 342897-54-5

Order Now

Specification Test Results

Melting point 76-78°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR