4-Ethoxy-2-phenyl-5-pyrimidinecarboxylic acid ≥95%
Supplier: Matrix Scientific
042863-500MG
042863-5G
101857-922EA
353.85
USD
101857-922
101857-926
4-Ethoxy-2-phenyl-5-pyrimidinecarboxylic acid ≥95%
4-Ethoxy-2-phenyl-5-pyrimidinecarboxylic acid
Hazard: Irritant
Formula:
C₁₃H₁₂N₂O₃ Melting Pt: 181…183 °C |
MDL Number:
MFCD06797376 CAS Number: 136326-10-8 |
Specification Test Results
Melting point | 181-183°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...