Pentafluorophenyl 4-ethylbenzenesulfonate ≥95%

Supplier: Matrix Scientific

042813-500MG 042813-1G 042813-5G
101857-674EA 375.38 USD
101857-674 101857-676 101857-678
Pentafluorophenyl 4-ethylbenzenesulfonate ≥95%
Pentafluorophenyl 4-ethylbenzenesulphonate

Hazard: Irritant

Formula: C₁₄H₉F₅O₃S
Melting Pt: 61…63 °C
MDL Number: MFCD05975134
CAS Number: 885950-41-4

Order Now

Specification Test Results

Melting point 61-63°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR