3-[(Dimethylamino)methyl]-4-hydroxy-5-methoxybenzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 3-((Dimethylamino)methyl)-4-hydroxy-5-methoxybenzaldehyde

042793-500MG 042793-1G 042793-5G
101857-558EA 181.54 USD
101857-558 101857-560 101857-562
3-[(Dimethylamino)methyl]-4-hydroxy-5-methoxybenzenecarbaldehyde ≥95%
3-[(Dimethylamino)methyl]-4-hydroxy-5-methoxybenzenecarbaldehyde

Hazard: Irritant

Formula: C₁₁H₁₅NO₃
Melting Pt: 140…141 °C
MDL Number: MFCD03011572
CAS Number: 87743-10-0

Order Now

Specification Test Results

Melting point 140-141°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR