Ethyl-8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate

042656-500MG 042656-1G
101856-836EA 278.46 USD
101856-836 101856-838
Ethyl-8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate ≥95%
Ethyl 8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate

Hazard: Irritant

Formula: C₁₁H₈ClF₃N₂O₂
MW: 292.65 g/mol
Melting Pt: 132…134 °C
MDL Number: MFCD01833013
CAS Number: 353258-31-8

Order Now

Specification Test Results

Melting point 132-134°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR