2-{[(2-Chlorophenyl)sulfonyl]amino}-3-phenylpropanoic acid ≥95%

Supplier: Matrix Scientific

042550-500MG 042550-1G 042550-5G
101856-306EA 181.54 USD
101856-306 101856-308 101856-310
2-{[(2-Chlorophenyl)sulfonyl]amino}-3-phenylpropanoic acid ≥95%
2-{[(2-Chlorophenyl)sulfonyl]amino}-3-phenylpropanoic acid

Hazard: Irritant

Formula: C₁₅H₁₄ClNO₄S
Melting Pt: 141…142 °C
MDL Number: MFCD00173004
CAS Number: 250714-62-6

Order Now

Specification Test Results

Melting point 141-142°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR