Boc-DL-6-methoxy-1,2,3,4-tetrahydronorharman-1-carboxylic acid

Supplier: Matrix Scientific

042134-250MG 042134-1G
101854-606EA 373.85 USD
101854-606 101854-608
Boc-DL-6-methoxy-1,2,3,4-tetrahydronorharman-1-carboxylic acid
Boc-DL-6-methoxy-1,2,3,4-tetrahydronorharman-1-carboxylic acid

Hazard: Irritant

Formula: C₁₈H₂₂N₂O₅
MDL Number: MFCD02682413
CAS Number: 1219423-55-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR