Fmoc-DL-thyronine

Supplier: Matrix Scientific

042001-1G 042001-5G
101854-050EA 1095.38 USD
101854-050 101854-052
Fmoc-DL-thyronine
Fmoc-DL-thyronine

Hazard: Irritant

Formula: C₃₀H₂₅NO₆
MDL Number: MFCD02682677
CAS Number: 1452575-06-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR