N-Boc-2-trifluoromethyl-L-phenylalanine
Supplier: Matrix Scientific
Synonyms:
2-(Trifluoromethyl)-L-phenylalanine, N-BOC protected
041827-1G
041827-5G
101853-362EA
1095.38
USD
101853-362
101853-364
N-Boc-2-trifluoromethyl-L-phenylalanine
N-Boc-2-trifluoromethyl-L-phenylalanine
Hazard: Irritant
Formula:
C₁₅H₁₈F₃NO₄ |
MDL Number:
MFCD01862946 CAS Number: 167993-21-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...