Boc-α-methyl-D-aspartic acid
Supplier: Matrix Scientific
041630-100MG
041630-500MG
101852-550EA
536.92
USD
101852-550
101852-552
Boc-α-methyl-D-aspartic acid
Boc-alpha-methyl-D-aspartic acid
Hazard: Irritant
Formula:
C₁₀H₁₇NO₆ |
MDL Number:
MFCD02682503 CAS Number: 1217826-44-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...