N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-cyclohexyl-L-alanine
Supplier: Matrix Scientific
Synonyms:
Fmoc-Cha-OH, N-Fmoc-3-cyclohexyl-L-alanine
041550-5G
041550-25G
101852-230EA
153.85
USD
101852-230
101852-232
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-cyclohexyl-L-alanine
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-cyclohexyl-L-alanine
CAS: 135673-97-1
MDL Number: MFCD00065614
Molecular Formula: C24H27NO4
Molecular Weight: 393,49
Hazard: Irritant
Formula:
C₂₄H₂₇NO₄ |
MDL Number:
MFCD00065614 CAS Number: 135673-97-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...