Fmoc-Dap(Alloc)-OH

Supplier: Matrix Scientific

Synonyms: Fmoc-Dap(Aloc)-OH, (S)-3-Allyloxycarbonylamino-2-(Fmoc-amino)propionic acid

041491-1G 041491-5G
101851-990EA 169.23 USD
101851-990 101851-992
Fmoc-Dap(Alloc)-OH
Fmoc-Dap(Alloc)-OH

Hazard: Irritant

Formula: C₂₂H₂₂N₂O₆
MDL Number: MFCD00273468
CAS Number: 188970-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR