Boc-D-Cha-OH
Supplier: Matrix Scientific
Synonyms:
Boc-β-cyclohexyl-D-Ala-OH
041439-1G
041439-5G
101851-782EA
21.54
USD
101851-782
101851-784
Boc-D-Cha-OH
Boc-D-Cha-OH
CAS: 127095-92-5
MDL Number: MFCD00133627
Molecular Formula: C14H25NO4
Molecular Weight: 271,36
Hazard: Irritant
Formula:
C₁₄H₂₅NO₄ MW: 271.357 g/mol Melting Pt: 64…67 °C |
MDL Number:
MFCD00133627 CAS Number: 127095-92-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...