Boc-Aib-OH

Supplier: Matrix Scientific

041421-25G
101851-712EA 44.62 USD
101851-712
Boc-Aib-OH
Boc-Aib-OH

Hazard: Irritant

Formula: C₉H₁₇NO₄
MDL Number: MFCD00042973
CAS Number: 30992-29-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR