(1R,3S)-Fmoc-3-aminocyclopentane-1-carboxylic acid

Supplier: Matrix Scientific

Synonyms: (1R,3S)-(-)-3-Aminocyclopentane-1-carboxylic acid, N-FMOC protected

041319-250MG 041319-1G
101851-326EA 300 USD
101851-326 101851-328
(1R,3S)-Fmoc-3-aminocyclopentane-1-carboxylic acid
(1R,3S)-Fmoc-3-aminocyclopentane-1-carboxylic acid

Hazard: Irritant

Formula: C₂₁H₂₁NO₄
MW: 351.41 g/mol
MDL Number: MFCD01311758
CAS Number: 220497-67-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR