Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-γ-trityl-L-asparagine
Supplier: Matrix Scientific
Synonyms:
Fmoc-Asn(Trt)-OH, Nɑ-Fmoc-Nγ-trityl-L-asparagine
041288-25G
101851-216EA
235.38
USD
101851-216
Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-γ-trityl-L-asparagine
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-gamma-trityl-L-asparagine
CAS: 132388-59-1
MDL Number: MFCD00077049
Molecular Formula: C38H32N2O5
Molecular Weight: 596,69
Hazard: Irritant
Formula:
C₃₈H₃₂N₂O₅ |
MDL Number:
MFCD00077049 CAS Number: 132388-59-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...