Fmoc-D-Arg(Mtr)-OH

Supplier: Matrix Scientific

Synonyms: Nα-Fmoc-Nomega-(4-methoxy-2,3,6-trimethylphenylsulfonyl)-D-arginine, Nα-Fmoc-Nomega-(4-methoxy-2,3,6-trimethylphenylsulphonyl)-D-arginine

041277-1G 041277-5G
101851-166EA 113.85 USD
101851-166 101851-168
Fmoc-D-Arg(Mtr)-OH
Fmoc-D-Arg(Mtr)-OH

CAS: 120075-24-3 MDL Number: MFCD00065616 Molecular Formula: C31H36N4O7S Molecular Weight: 608,72 Hazard: Irritant

Formula: C₃₁H₃₆N₄O₇S
MW: 608.71 g/mol
MDL Number: MFCD00065616
CAS Number: 120075-24-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR