1-Boc-4-bromo-3-cyanoacetylindole

Supplier: Matrix Scientific

041131-250MG
101850-866EA 558.46 USD
101850-866
1-Boc-4-bromo-3-cyanoacetylindole
1-Boc-4-bromo-3-cyanoacetylindole

Hazard: Irritant

Formula: C₁₆H₁₅BrN₂O₃
MDL Number: MFCD11849918
CAS Number: 1171917-34-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR