1-[3-(Chloromethyl)phenyl]-3,5-dimethyl-1H-pyrazole hydrochloride

Supplier: Matrix Scientific

040981-500MG
101850-548EA 146.15 USD
101850-548
1-[3-(Chloromethyl)phenyl]-3,5-dimethyl-1H-pyrazole hydrochloride
1-[3-(Chloromethyl)phenyl]-3,5-dimethyl-1H-pyrazole hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₄Cl₂N₂
MDL Number: MFCD12028461
CAS Number: 1185298-82-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR