4-[(2-Methylbutanoyl)amino]benzoic acid

Supplier: Matrix Scientific

040901-500MG
101850-396EA 193.85 USD
101850-396
4-[(2-Methylbutanoyl)amino]benzoic acid
4-[(2-Methylbutanoyl)amino]benzoic acid

Hazard: Irritant

Formula: C₁₂H₁₅NO₃
MDL Number: MFCD08723294
CAS Number: 925068-68-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR