A1c-Cellular® Linearity Control, Streck
Supplier: STRECK LABORATORIES
285536
10185-902EA 703.96 USD
10185-902
A1c-Cellular® Linearity Control, Streck
Controls and Calibration Verification Materials
A linearity product with instrument-specific target values and ranges for immediate results.
  • Ready-to-use liquid control
  • For immunoassay and ionic exchange HPLC methodologies
  • Assayed on top 10 HbA1c analyzers
  • Tests the entire HbA1c procedure, including the lysing of red blood cells
  • Available in a 5-level set
Available in plastic cap-pierceable vials for analyzer autosampling. Only available HbA1c linearity material containing intact red blood cells.