Merit™ Simpson Uterine Sound, Physician Grade, Sklar
Supplier: Sklar
98-382
10185-322EA 55.59 USD
10185-322
Merit™ Simpson Uterine Sound, Physician Grade, Sklar
Cannulae
The Merit™ Simpson Uterine Sound is an office-grade, malleable, silver plated OB/GYN instrument used for probing and dilating the uterus, as well as measuring the length of the cervical canal.
  • It is round and thin with a small nodule at the tip and a thumb grip handle
  • This product has a gently curved tip, graduated centimeter markings, and an overall length of 121/2"
It is very similar to the Sims uterine sound.