{[4-(4-Fluorophenyl)-6-(trifluoromethyl)pyrimidin-2-yl]thio}acetic acid

Supplier: Matrix Scientific

040694-500MG
101849-990EA 290.77 USD
101849-990
{[4-(4-Fluorophenyl)-6-(trifluoromethyl)pyrimidin-2-yl]thio}acetic acid
{[4-(4-Fluorophenyl)-6-(trifluoromethyl)pyrimidin-2-yl]thio}acetic acid

Hazard: Irritant

Formula: C₁₃H₈F₄N₂O₂S
MDL Number: MFCD02735802
CAS Number: 505049-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR