2-Methyl-N-phenyl-N-(2,3,4,5-tetrahydro-1,4-benzoxazepin-7-ylmethyl)propanamide hydrochloride

Supplier: Matrix Scientific

040650-500MG
101849-908EA 290.77 USD
101849-908
2-Methyl-N-phenyl-N-(2,3,4,5-tetrahydro-1,4-benzoxazepin-7-ylmethyl)propanamide hydrochloride
2-Methyl-N-phenyl-N-(2,3,4,5-tetrahydro-1,4-benzoxazepin-7-ylmethyl)propanamide hydrochloride

Hazard: Irritant

Formula: C₂₀H₂₅ClN₂O₂
MDL Number: MFCD12028356
CAS Number: 1185302-76-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR