5-(1-Ethylpropyl)-1,3,4-thiadiazol-2-amine

Supplier: Matrix Scientific

040380-500MG
101849-396EA 193.85 USD
101849-396
5-(1-Ethylpropyl)-1,3,4-thiadiazol-2-amine
5-(1-Ethylpropyl)-1,3,4-thiadiazol-2-amine

Hazard: Irritant

Formula: C₇H₁₃N₃S
MW: 171.27 g/mol
MDL Number: MFCD01131203
CAS Number: 229003-14-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR