5-(Methoxymethyl)-1,3,4-thiadiazol-2-amine

Supplier: Matrix Scientific

040377-500MG
101849-390EA 127.69 USD
101849-390
5-(Methoxymethyl)-1,3,4-thiadiazol-2-amine
5-(Methoxymethyl)-1,3,4-thiadiazol-2-amine

Hazard: Irritant

Formula: C₄H₇N₃OS
MDL Number: MFCD01203002
CAS Number: 15884-86-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR