(4-Chloro-1H-pyrazol-1-yl)acetaldehyde

Supplier: Matrix Scientific

040238-500MG
101849-114EA 484.62 USD
101849-114
(4-Chloro-1H-pyrazol-1-yl)acetaldehyde
(4-Chloro-1H-pyrazol-1-yl)acetaldehyde

Hazard: Irritant

Formula: C₅H₅ClN₂O
MW: 144.56 g/mol
MDL Number: MFCD04969646
CAS Number: 1015939-57-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR