(5E)-5-(4-Hydroxy-3-methoxybenzylidene)rhodanine

Supplier: Matrix Scientific

040196-500MG
101849-030EA 116.92 USD
101849-030
(5E)-5-(4-Hydroxy-3-methoxybenzylidene)rhodanine
(5E)-5-(4-Hydroxy-3-methoxybenzylidene)rhodanine

Hazard: Irritant

Formula: C₁₁H₉NO₃S₂
CAS Number: 5447-37-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR