(4E)-3-(Chloromethyl)-4-(4-fluorobenzylidene)isoxazol-5(4H)-one

Supplier: Matrix Scientific

039310-500MG
101847-236EA 193.85 USD
101847-236
(4E)-3-(Chloromethyl)-4-(4-fluorobenzylidene)isoxazol-5(4H)-one
(4E)-3-(Chloromethyl)-4-(4-fluorobenzylidene)isoxazol-5(4H)-one

Hazard: Irritant

Formula: C₁₁H₇ClFNO₂
MDL Number: MFCD12027797
CAS Number: 1142199-23-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR