{(4-Methoxyphenyl)[2-(2-methylpiperidin-1-yl)-2-oxoethyl]amino}acetic acid

Supplier: Matrix Scientific

039175-500MG
101846-970EA 193.85 USD
101846-970
{(4-Methoxyphenyl)[2-(2-methylpiperidin-1-yl)-2-oxoethyl]amino}acetic acid
{(4-Methoxyphenyl)[2-(2-methylpiperidin-1-yl)-2-oxoethyl]amino}acetic acid

Hazard: Irritant

Formula: C₁₇H₂₄N₂O₄
MDL Number: MFCD12027679
CAS Number: 1142205-83-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR