(2E)-1-(4-Aminophenyl)-3-(3-chlorophenyl)prop-2-en-1-one

Supplier: Matrix Scientific

038984-500MG
101846-592EA 193.85 USD
101846-592
(2E)-1-(4-Aminophenyl)-3-(3-chlorophenyl)prop-2-en-1-one
(2E)-1-(4-Aminophenyl)-3-(3-chlorophenyl)prop-2-en-1-one

Hazard: Irritant

Formula: C₁₅H₁₂ClNO
MDL Number: MFCD09559384
CAS Number: 952578-23-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR