(2E)-1-(4-Aminophenyl)-3-(2-chloro-6-fluorophenyl)prop-2-en-1-one

Supplier: Matrix Scientific

038982-500MG
101846-588EA 193.85 USD
101846-588
(2E)-1-(4-Aminophenyl)-3-(2-chloro-6-fluorophenyl)prop-2-en-1-one
(2E)-1-(4-Aminophenyl)-3-(2-chloro-6-fluorophenyl)prop-2-en-1-one

Hazard: Irritant

Formula: C₁₅H₁₁ClFNO
MDL Number: MFCD09559382
CAS Number: 682745-99-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR