2-Chloro-1-(2,5-dimethyl-1H-indol-3-yl)ethanone

Supplier: Matrix Scientific

038850-500MG
101846-332EA 290.77 USD
101846-332
2-Chloro-1-(2,5-dimethyl-1H-indol-3-yl)ethanone
2-Chloro-1-(2,5-dimethyl-1H-indol-3-yl)ethanone

Hazard: Irritant

Formula: C₁₂H₁₂ClNO
MDL Number: MFCD11589635
CAS Number: 887569-01-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR