Ethyl-3-phenyl-1H-indole-2-carboxylate

Supplier: Matrix Scientific

038781-500MG
101846-198EA 216.92 USD
101846-198
Ethyl-3-phenyl-1H-indole-2-carboxylate
Ethyl-3-phenyl-1H-indole-2-carboxylate

Hazard: Irritant

Formula: C₁₇H₁₅NO₂
MW: 265.31 g/mol
Melting Pt: 134…136 °C
MDL Number: MFCD00974211
CAS Number: 37129-23-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR