Methyl-4-ethoxy-1H-indole-2-carboxylate

Supplier: Matrix Scientific

038761-500MG
101846-158EA 290.77 USD
101846-158
Methyl-4-ethoxy-1H-indole-2-carboxylate
Methyl-4-ethoxy-1H-indole-2-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₃NO₃
MW: 219.24 g/mol
MDL Number: MFCD04967029
CAS Number: 185212-25-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR