4-Chloro-3-(2-mercapto-4,4-dimethylpyrimidin-1(4H)-yl)benzoic acid

Supplier: Matrix Scientific

038750-500MG
101846-136EA 193.85 USD
101846-136
4-Chloro-3-(2-mercapto-4,4-dimethylpyrimidin-1(4H)-yl)benzoic acid
4-Chloro-3-(2-mercapto-4,4-dimethylpyrimidin-1(4H)-yl)benzoic acid

Hazard: Irritant

Formula: C₁₃H₁₃ClN₂O₂S
MW: 296.78 g/mol
MDL Number: MFCD12027437
CAS Number: 1142213-03-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR