4,4-Dimethyl-1-(2-methyl-5-nitrophenyl)-1,4-dihydropyrimidine-2-thiol

Supplier: Matrix Scientific

038744-500MG
101846-124EA 193.85 USD
101846-124
4,4-Dimethyl-1-(2-methyl-5-nitrophenyl)-1,4-dihydropyrimidine-2-thiol
4,4-Dimethyl-1-(2-methyl-5-nitrophenyl)-1,4-dihydropyrimidine-2-thiol

Hazard: Irritant

Formula: C₁₃H₁₅N₃O₂S
MDL Number: MFCD12027431
CAS Number: 1142212-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR