3-Chloro-6-[4-(piperidin-1-ylcarbonyl)piperidin-1-yl]pyridazine

Supplier: Matrix Scientific

038717-500MG
101846-070EA 193.85 USD
101846-070
3-Chloro-6-[4-(piperidin-1-ylcarbonyl)piperidin-1-yl]pyridazine
3-Chloro-6-[4-(piperidin-1-ylcarbonyl)piperidin-1-yl]pyridazine

Hazard: Irritant

Formula: C₁₅H₂₁ClN₄O
MDL Number: MFCD12027416
CAS Number: 1142214-17-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR