4-(Trifluoromethyl)phenyl isocyanate ≥95%

Supplier: Matrix Scientific
Danger

038715-25G 038715-100G
101846-064EA 230.77 USD
101846-064 101846-066
4-(Trifluoromethyl)phenyl isocyanate ≥95%
4-(Trifluoromethyl)phenyl isocyanate
Formula: C₈H₄F₃NO
Boiling Pt: 132 °C (20mm)
Melting Pt: 29…31 °C
MDL Number: MFCD00002028
CAS Number: 1548-13-6
UN: 2923
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR