4-Fluorobenzyl isocyanate

Supplier: Matrix Scientific

038686-500MG
101846-006EA 290.77 USD
101846-006
4-Fluorobenzyl isocyanate
4-Fluorobenzyl isocyanate

Hazard: Irritant

Formula: C₈H₆FNO
MDL Number: MFCD00673062
CAS Number: 132740-43-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR